3,5-Dimethyl-1,2,4-triazole ≥95.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3,5-dimethyl-4H-1,2,4-triazole
D4481-1G
D4481-5G
TCIAD4481-1GEA
30
EUR
TCIAD4481-1G
TCIAD4481-5G
3,5-Dimethyl-1,2,4-triazole ≥95.0% (by GC, titration analysis)
3,5-Dimethyl-1,2,4-triazole
Formula:
C₄H₇N₃ Peso molecolare: 97,12 g/mol Punto di ebollizione: 260°C |
MDL Number:
MFCD00656686 CAS Number: 7343-34-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Melting point | 139.0 to 144.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...