Stampa…

4,4'-Dimethyl-2,2'-bipyridyl 1-Oxide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4,4'-Dimethyl-[2,2'-bipyridine] 1-oxide, 4,4'-Dimethyl-2,2'-bipyridine 1-Oxide

D4467-1G D4467-5G
TCIAD4467-1GEA 66 EUR
TCIAD4467-1G TCIAD4467-5G
4,4'-Dimethyl-2,2'-bipyridyl 1-Oxide ≥98.0% (by GC)
4,4'-Dimethyl-2,2'-bipyridyl 1-Oxide
Formula: C₁₂H₁₂N₂O
Peso molecolare: 200,24 g/mol
MDL Number: MFCD11042487
CAS Number: 81998-03-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 89.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR