Stampa…

3,6-Diphenylcarbazole ≥99.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,6-Diphenyl-9H-carbazole

D4433-1G D4433-5G
TCIAD4433-1GEA 27 EUR
TCIAD4433-1G TCIAD4433-5G
3,6-Diphenylcarbazole ≥99.0% (by GC)
3,6-Diphenylcarbazole
Formula: C₂₄H₁₇N
Peso molecolare: 319,41 g/mol
MDL Number: MFCD00222619
CAS Number: 56525-79-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 182.0 to 186.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR