Stampa…

Dimethyl-4-nitrophthalate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Dimethyl 4-nitrophthalate

D4424-1G D4424-5G
TCIAD4424-1GEA 22 EUR
TCIAD4424-1G TCIAD4424-5G
Dimethyl-4-nitrophthalate ≥98.0% (by GC)
Dimethyl-4-nitrophthalate
Formula: C₁₀H₉NO₆
Peso molecolare: 239,19 g/mol
Punto di ebollizione: 195°C
MDL Number: MFCD00017191
CAS Number: 610-22-0
EINECS: 210-212-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 69.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR