Stampa…

Dimethyl (2-oxo-3-phenoxypropyl)phosphonate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Dimethyl (3-Phenoxyacetonyl)phosphonate, (2-Oxo-3-phenoxypropyl)phosphonic Acid Dimethyl Ester, (3-Phenoxyacetonyl)phosphonic Acid Dimethyl Ester

D4408-200MG D4408-1G
TCIAD4408-200MGEA 64 EUR
TCIAD4408-200MG TCIAD4408-1G
Dimethyl (2-oxo-3-phenoxypropyl)phosphonate ≥98.0% (by GC)
Dimethyl (2-oxo-3-phenoxypropyl)phosphonate
Formula: C₁₁H₁₅O₅P
Peso molecolare: 258,21 g/mol
Punto di ebollizione: 175°C
MDL Number: MFCD00010225
CAS Number: 40665-68-7

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR