4',7-Dimethoxyisoflavone ≥97.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
7-Methoxy-3-(4-methoxyphenyl)chromen-4-one, 7-Methoxy-3-(4-methoxyphenyl)-4-benzopyrone, 4',7-Dimethoxyisoflavone
D4389-1G
D4389-5G
TCIAD4389-1GEA
92
EUR
TCIAD4389-1G
TCIAD4389-5G
4',7-Dimethoxyisoflavone ≥97.0% (by GC)
4',7-Dimethoxyisoflavone
Formula:
C₁₇H₁₄O₄ Peso molecolare: 282,3 g/mol |
MDL Number:
MFCD00075889 CAS Number: 1157-39-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 161.0 to 165.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...