Stampa…

1,2-Bis(1-naphthyl)ethane ≥94.0% (by GC)

Fornitore: TCI

D4371-1G D4371-5G
TCIAD4371-1GEA 55 EUR
TCIAD4371-1G TCIAD4371-5G
1,2-Bis(1-naphthyl)ethane ≥94.0% (by GC)
1,2-Bis(1-naphthyl)ethane
Formula: C₂₂H₁₈
MDL Number: MFCD00037736
CAS Number: 15374-45-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 94.0 %
Melting point 159.0 to 163.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR