2,4-Dihydroxy-6-methylpyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Methyl-2,4-pyridinediol, 4,6-Dihydroxy-2-picoline, 4-Hydroxy-6-methyl-2-pyridone, 4-Hydroxy-6-methylpyridin-2(1H)-one
D4361-1G
D4361-5G
TCIAD4361-1GEA
24
EUR
TCIAD4361-1G
TCIAD4361-5G
2,4-Dihydroxy-6-methylpyridine ≥98.0% (by GC, titration analysis)
2,4-Dihydroxy-6-methylpyridine
Formula:
C₆H₇NO₂ Peso molecolare: 125,13 g/mol Punto di fusione: 330°C |
MDL Number:
MFCD01860849 CAS Number: 3749-51-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...