2,6-Dimethylphenoxyacetic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-(2,6-Dimethylphenoxy)acetic acid
D4345-1G
D4345-5G
TCIAD4345-1GEA
34
EUR
TCIAD4345-1G
TCIAD4345-5G
2,6-Dimethylphenoxyacetic acid ≥98.0% (by GC, titration analysis)
2,6-Dimethylphenoxyacetic acid
Formula:
C₁₀H₁₂O₃ Peso molecolare: 180,20 g/mol |
MDL Number:
MFCD00156912 CAS Number: 13335-71-2 EINECS: 430-910-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 137.0 to 141.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...