Stampa…

2,6-Dibromonaphthalene-1,4,5,8-tetracarboxylic dianhydride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

D4339-5G D4339-1G
TCIAD4339-5GEA 627 EUR
TCIAD4339-5G TCIAD4339-1G
2,6-Dibromonaphthalene-1,4,5,8-tetracarboxylic dianhydride ≥98.0% (by HPLC, titration analysis)
2,6-Dibromonaphthalene-1,4,5,8-tetracarboxylic dianhydride
Formula: C₁₄H₂Br₂O₆
Peso molecolare: 425,97 g/mol
Punto di fusione: 350°C
MDL Number: MFCD00406909
CAS Number: 83204-68-6

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR