Stampa…
4,4'-Dibromo-1,1'-spirobiindane-7,7'-diol ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms: 4,4'-Dibromo-7,7'-dihydroxy-1,1'-spirobiindane
D4332-5G D4332-1G
TCIAD4332-5GEA 496 EUR
TCIAD4332-5G TCIAD4332-1G
4,4'-Dibromo-1,1'-spirobiindane-7,7'-diol ≥98.0% (by GC)
4,4'-Dibromo-1,1'-spirobiindane-7,7'-diol
Formula: C₁₇H₁₄Br₂O₂
Peso molecolare: 410,11 g/mol
MDL Number: MFCD16660947
CAS Number: 681481-91-4
Visualizza i codici
Purity (GC) min. 98.0 %
Melting point 146.0 to 150.0 deg-C