Stampa…

9,9-Dihexylfluorene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 9,9-Dihexyl-9H-fluorene

D4327-1G D4327-5G
TCIAD4327-1GEA 88 EUR
TCIAD4327-1G TCIAD4327-5G
9,9-Dihexylfluorene ≥98.0% (by GC)
9,9-Dihexylfluorene
Formula: C₂₅H₃₄
Peso molecolare: 334,55 g/mol
Punto di fusione: 32°C
MDL Number: MFCD14584648
CAS Number: 123863-97-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR