Stampa…

3,5-Dibromopyridine-N-oxide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,5-Dibromopyridine1-oxide

D4326-1G D4326-5G
TCIAD4326-1GEA 33 EUR
TCIAD4326-1G TCIAD4326-5G
3,5-Dibromopyridine-N-oxide ≥98.0% (by GC)
3,5-Dibromopyridine-N-oxide
Formula: C₅H₃Br₂NO
Peso molecolare: 252,8914 g/mol
MDL Number: MFCD00160750
CAS Number: 2402-99-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 143.0 to 146.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR