Stampa…

1,11-Dioxa[11]paracyclophane ≥97.0% (by GC)

Fornitore: TCI EUROPE

D4298-250MG D4298-1G
TCIAD4298-250MGEA 70 EUR
TCIAD4298-250MG TCIAD4298-1G
1,11-Dioxa[11]paracyclophane ≥97.0% (by GC)
1,11-Dioxa[11]paracyclophane
Formula: C₁₅H₂₂O₂
Peso molecolare: 234,34 g/mol
CAS Number: 6571-51-3

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 97.0 %
Melting point 53.0 to 57.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR