1,3-Dibromo-2-iodobenzene ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,6-Dibromoiodobenzene
D4294-1G
D4294-5G
TCIAD4294-1GEA
69
EUR
TCIAD4294-1G
TCIAD4294-5G
1,3-Dibromo-2-iodobenzene ≥98.0% (by GC)
1,3-Dibromo-2-iodobenzene
Formula:
C₆H₃Br₂I Peso molecolare: 361,80 g/mol |
MDL Number:
MFCD00192666 CAS Number: 19821-80-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...