(2,5-Dimethylphenyl)acetonitrile ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
D4271-5G
D4271-25G
TCIAD4271-5GEA
27
EUR
TCIAD4271-5G
TCIAD4271-25G
(2,5-Dimethylphenyl)acetonitrile ≥98.0% (by GC)
(2,5-Dimethylphenyl)acetonitrile
Formula:
C₁₀H₁₁N Punto di ebollizione: 79°C |
MDL Number:
MFCD00013816 CAS Number: 16213-85-7 EINECS: 240-339-3 UN: 3439 ADR: 6.1,III |
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |