Stampa…

N-[3-[3-(Dimethylamino)acryloyl]phenyl]-N-ethylacetamide ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: N-(3-(3-(Dimethylamino)acryloyl)phenyl)-N-ethylacetamide

D4259-1G D4259-5G
TCIAD4259-1GEA 16 EUR
TCIAD4259-1G TCIAD4259-5G
N-[3-[3-(Dimethylamino)acryloyl]phenyl]-N-ethylacetamide ≥98.0% (by GC, titration analysis)
N-[3-[3-(Dimethylamino)acryloyl]phenyl]-N-ethylacetamide
Formula: C₁₅H₂₀N₂O₂
Peso molecolare: 260,34 g/mol
MDL Number: MFCD04117958
CAS Number: 96605-66-2

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 111.0 to 115.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR