Stampa…

Droxidopa ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (2S,3R)-2-Amino-3-(3,4-dihydroxyphenyl)-3-hydroxypropionic Acid, L-threo-3,4-Dihydroxyphenylserine

D4235-50MG D4235-200MG
TCIAD4235-50MGEA 100 EUR
TCIAD4235-50MG TCIAD4235-200MG
Droxidopa ≥98.0% (by HPLC, titration analysis)
Droxidopa
Formula: C₉H₁₁NO₅
Peso molecolare: 213,19 g/mol
Punto di fusione: 225°C
MDL Number: MFCD00799030
CAS Number: 23651-95-8
Merck Index: 14,03457

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -38.0 to -43.0 deg (C=0.5、0.1mol/L HCl) (after drying)
Drying loss max. 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR