Stampa…

4,4'-Diethynylbiphenyl ≥98.0% (by GC)

Fornitore: TCI

Synonyms: 4,4'-Diethynyl-1,1'-biphenyl

D4233-200MG D4233-1G
TCIAD4233-200MGEA 82 EUR
TCIAD4233-200MG TCIAD4233-1G
4,4'-Diethynylbiphenyl ≥98.0% (by GC)
4,4'-Diethynylbiphenyl
Formula: C₁₆H₁₀
MDL Number: MFCD00458964
CAS Number: 38215-38-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 166.0 to 170.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR