1,8-Dibromoanthracene ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₄H₈Br₂ |
CAS Number:
131276-24-9 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 166.0 to 170.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...