Stampa…

Duloxetine hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (S)-(+)-N-Methyl-3-(1-naphthyloxy)-3-(2-thienyl)propylamine Hydrochloride, (S)-Duloxetine hydrochloride

D4223-1G D4223-5G
TCIAD4223-1GEA 71 EUR
TCIAD4223-1G TCIAD4223-5G
Duloxetine hydrochloride ≥98.0% (by HPLC, titration analysis)
Duloxetine hydrochloride
Formula: C₁₈H₁₉NOS
Peso molecolare: 333,87 g/mol
MDL Number: MFCD06407958
CAS Number: 136434-34-9
Merck Index: 14,03465

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 162.0 to 166.0 deg-C
Specific rotation [a]20/D +120 to +124 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR