Stampa…

4,5-Dihydro-6-methyl-3(2H)-pyridazinone ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 6-Methyl-4,5-dihydropyridazin-3(2H)-one

D4216-5G D4216-25G
TCIAD4216-5GEA 16 EUR
TCIAD4216-5G TCIAD4216-25G
4,5-Dihydro-6-methyl-3(2H)-pyridazinone ≥98.0% (by GC)
4,5-Dihydro-6-methyl-3(2H)-pyridazinone
Formula: C₅H₈N₂O
MDL Number: MFCD00006455
CAS Number: 5157-08-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6a8b8acd78c471e426d78958852f68b1fb16f3a&vid=120acd6187aa1ccb823f3207885826c081c60c74&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR