Stampa…

4,4'-Di(4-pyridyl)biphenyl ≥95.0% (by GC)

Fornitore: TCI

Synonyms: 4,4'-Bis(4-pyridyl)biphenyl, 4,4'-Di(pyridin-4-yl)-1,1'-biphenyl

D4203-200MG D4203-1G
TCIAD4203-200MGEA 77 EUR
TCIAD4203-200MG TCIAD4203-1G
4,4'-Di(4-pyridyl)biphenyl ≥95.0% (by GC)
4,4'-Di(4-pyridyl)biphenyl
Formula: C₂₂H₁₆N₂
Peso molecolare: 308.38 g/mol
Punto di fusione: 308°C
MDL Number: MFCD11977635
CAS Number: 319430-87-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR