Stampa…

3,5-Diphenyl-1H-pyrazole ≥98.0% (by GC)

Fornitore: TCI

D4197-5G D4197-25G
TCIAD4197-5GEA 40 EUR
TCIAD4197-5G TCIAD4197-25G
3,5-Diphenyl-1H-pyrazole ≥98.0% (by GC)
3,5-Diphenyl-1H-pyrazole
Formula: C₁₅H₁₂N₂
MDL Number: MFCD00039675
CAS Number: 1145-01-3
EINECS: 214-543-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 200.0 to 204.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR