2,7-Di-tert-butylpyrene ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₂₄H₂₆ |
MDL Number:
MFCD01013641 CAS Number: 24300-91-2 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 207.0 to 211.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...