4,4'-Dibromo-2,2'-bithiophene ≥98.0% (by GC)
Fornitore: TCI EUROPE
D4181-1G
D4181-5G
TCIAD4181-1GEA
159
EUR
TCIAD4181-1G
TCIAD4181-5G
4,4'-Dibromo-2,2'-bithiophene ≥98.0% (by GC)
4,4'-Dibromo-2,2'-bithiophene
Formula:
C₈H₄Br₂S₂ Peso molecolare: 324,05 g/mol |
MDL Number:
MFCD00508561 CAS Number: 51285-60-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 132.0 to 136.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...