Stampa…

trans-3,4-Difluorocinnamic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,4-Difluorocinnamic acid

D4161-1G D4161-5G
TCIAD4161-1GEA 17 EUR
TCIAD4161-1G TCIAD4161-5G
trans-3,4-Difluorocinnamic acid ≥98.0% (by GC, titration analysis)
trans-3,4-Difluorocinnamic acid
Formula: C₉H₆F₂O₂
Peso molecolare: 184,14 g/mol
MDL Number: MFCD00010320
CAS Number: 112897-97-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 194.0 to 198.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR