2-((Diphenylmethylene)amino)acetonitrile ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
N-(Diphenylmethylene)aminoacetonitrile
Formula:
C₁₅H₁₂N₂ Peso molecolare: 220.2700 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00009970 CAS Number: 70591-20-7 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...