Stampa…

4,6-Dichloro-2-(methylsulphonyl)pyrimidine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4,6-Dichloro-2-(methanesulphonyl)pyrimidine, 4,6-Dichloro-2-methylsulphonylpyrimidine

D4135-1G D4135-5G
TCIAD4135-1GEA 25 EUR
TCIAD4135-1G TCIAD4135-5G
4,6-Dichloro-2-(methylsulphonyl)pyrimidine ≥98.0% (by GC)
4,6-Dichloro-2-(methylsulphonyl)pyrimidine
Formula: C₅H₄Cl₂N₂O₂S
Peso molecolare: 227,06 g/mol
MDL Number: MFCD00672147
CAS Number: 4489-34-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 118.0 to 122.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR