2-Amino-3,4-dimethylbenzoic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
D4120-5G
D4120-25G
TCIAD4120-5GEA
20
EUR
TCIAD4120-5G
TCIAD4120-25G
2-Amino-3,4-dimethylbenzoic acid ≥98.0% (by HPLC, titration analysis)
2-Amino-3,4-dimethylbenzoic acid
Formula:
C₉H₁₁NO₂ Peso molecolare: 165,19 g/mol |
MDL Number:
MFCD00130041 CAS Number: 50419-58-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...