3,5-Dichloropyridine-2-carbonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
3,5-Dichloropicolinonitrile, 3,5-Dichloro-2-cyanopyridine
D4059-1G
D4059-5G
TCIAD4059-1GEA
18
EUR
TCIAD4059-1G
TCIAD4059-5G
3,5-Dichloropyridine-2-carbonitrile ≥98.0% (by GC)
3,5-Dichloropyridine-2-carbonitrile
Formula:
C₆H₂Cl₂N₂ Peso molecolare: 173,00 g/mol |
MDL Number:
MFCD03788758 CAS Number: 85331-33-5 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 100.0 to 104.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...