Stampa…

9,9-Dimethylxanthene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 9,9-Dimethyl-9H-xanthene

D4055-5G D4055-1G
TCIAD4055-5GEA 78 EUR
TCIAD4055-5G TCIAD4055-1G
9,9-Dimethylxanthene ≥98.0% (by GC)
9,9-Dimethylxanthene
Formula: C₁₅H₁₄O
Peso molecolare: 210,28 g/mol
Punto di ebollizione: 115°C
MDL Number: MFCD00134434
CAS Number: 19814-75-6

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Yellow to Green powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR