Stampa…
5,5''-Dibromo-2,2':5',2''-terthiophene ≥97.0% (by GC)
Fornitore: TCI EUROPE
Synonyms: 2,5-Bis(5-bromo-2-thienyl)thiophene
D4050-1G D4050-5G
TCIAD4050-1GEA 141 EUR
TCIAD4050-1G TCIAD4050-5G
5,5''-Dibromo-2,2':5',2''-terthiophene ≥97.0% (by GC)
5,5''-Dibromo-2,2':5',2''-terthiophene
Formula: C₁₂H₆Br₂S₃
Peso molecolare: 406,17 g/mol
MDL Number: MFCD00219109
CAS Number: 98057-08-0
Visualizza i codici
Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 97.0 %
Melting point 156.0 to 160.0 deg-C