3,5-Difluoropyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
D4038-1G
D4038-5G
TCIAD4038-1GEA
26
EUR
TCIAD4038-1G
TCIAD4038-5G
3,5-Difluoropyridine ≥98.0% (by GC)
3,5-Difluoropyridine
Formula:
C₅H₃F₂N Peso molecolare: 115,08 g/mol Punto di ebollizione: 93°C Flash Pt: 7°C |
MDL Number:
MFCD03412222 CAS Number: 71902-33-5 UN: 1993 ADR: 3,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...