Stampa…

Diethyl-3,5-dimethyl-2,4-pyrroledicarboxylate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 3,5-Dimethyl-2,4-pyrroledicarboxylic Acid Diethyl Ester, Diethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate

D4012-25G D4012-5G
TCIAD4012-25GEA 348 EUR
TCIAD4012-25G TCIAD4012-5G
Diethyl-3,5-dimethyl-2,4-pyrroledicarboxylate ≥98.0% (by GC)
Diethyl-3,5-dimethyl-2,4-pyrroledicarboxylate
Formula: C₁₂H₁₇NO₄
Peso molecolare: 239,27 g/mol
MDL Number: MFCD00005218
CAS Number: 2436-79-5
EINECS: 219-431-2

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 134.0 to 138.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR