2,4-Dihydroxy-3-nitropyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Nitropyridine-2,4-diol
D4002-25G
D4002-5G
TCIAD4002-25GEA
155
EUR
TCIAD4002-25G
TCIAD4002-5G
2,4-Dihydroxy-3-nitropyridine ≥98.0% (by GC)
2,4-Dihydroxy-3-nitropyridine
Formula:
C₅H₄N₂O₄ Peso molecolare: 156,10 g/mol Punto di fusione: 264°C |
MDL Number:
MFCD01075671 CAS Number: 89282-12-2 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...