Stampa…

Daphnetin ≥90.0% (by HPLC)

Fornitore: TCI
Attenzione

Synonyms: 7,8-Dihydroxy-2H-chromen-2-one, 7,8-Dihydroxycoumarin

D4001-1G D4001-5G
TCIAD4001-1GEA 138 EUR
TCIAD4001-1G TCIAD4001-5G
Daphnetin ≥90.0% (by HPLC)
Daphnetin
Formula: C₉H₆O₄
Punto di fusione: 256°C
MDL Number: MFCD00016977
CAS Number: 486-35-1
EINECS: 207-632-8
Merck Index: 14,02817

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 90.0 area%
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR