9,10-Di(1-naphthyl)anthracene ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
9,10-Di(naphthalen-1-yl)anthracene
D3975-1G
D3975-5G
TCIAD3975-1GEA
76
EUR
TCIAD3975-1G
TCIAD3975-5G
9,10-Di(1-naphthyl)anthracene ≥98.0% (by HPLC)
9,10-Di(1-naphthyl)anthracene
Formula:
C₃₄H₂₂ Peso molecolare: 430,55 g/mol Punto di fusione: 360°C |
MDL Number:
MFCD04222155 CAS Number: 26979-27-1 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% (total of isomer) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...