4-(Dimethylamino)butyraldehyde diethyl acetal ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4,4-Diethoxy-N,N-dimethylbutylamine, 4,4-Diethoxy-N,N-dimethyl-1-butanamine
D3973-25G
D3973-5G
TCIAD3973-25GEA
69
EUR
TCIAD3973-25G
TCIAD3973-5G
4-(Dimethylamino)butyraldehyde diethyl acetal ≥98.0% (by GC, titration analysis)
4-(Dimethylamino)butyraldehyde diethyl acetal
Formula:
C₁₀H₂₃NO₂ Peso molecolare: 189,30 g/mol Punto di ebollizione: 95°C Flash Pt: 70°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00671479 CAS Number: 1116-77-4 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...