3,6-Di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione ≥95.0% (by HPLC, total nitrogen)
Fornitore: TCI
Formula:
C₁₄H₈N₂O₂S₂ |
MDL Number:
MFCD20257907 CAS Number: 850583-75-4 |
Risultati del test di specificazione
Appearance | Dark green to Dark red to Black powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (with Total Nitrogen) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...