Stampa…

Diethyl-2,5-dibromoadipate ≥98.0% (by GC)

Fornitore: TCI
Pericolo

D3965-25G D3965-5G
TCIAD3965-25GEA 84 EUR
TCIAD3965-25G TCIAD3965-5G
Diethyl-2,5-dibromoadipate ≥98.0% (by GC)
Diethyl-2,5-dibromoadipate
Formula: C₁₀H₁₆Br₂O₄
Punto di ebollizione: 134°C
MDL Number: MFCD00075379
CAS Number: 869-10-3
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63303a8e597951722d12230d0389e1541726c6ac&vid=14d8fbc0d5340a9e80633c74812145bd5b8dd1fd&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 68.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR