Stampa…

Sodio β-glicerofosfato pentaidrato ≥95.0% (by titrimetric analysis) for biochemical research

Fornitore: TCI

D3964-1G D3964-5G
TCIAD3964-1GEA 22 EUR
TCIAD3964-1G TCIAD3964-5G
Sodio β-glicerofosfato pentaidrato ≥95.0% (by titrimetric analysis) for biochemical research
Sodio β-glicerofosfato pentaidrato
Formula: C₃H₇Na₂O₆P·5H₂O
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00002135
CAS Number: 13408-09-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=caae63c2e0a5b37ea2a369c5ab1b964c8aa89553&vid=d59a1c068e20c9a763710440aa58dadd2156808c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White powder to crystal
Purity (Neutralization titration) min. 95.0 % (calcd.on anh.substance)
Water 27.0 to 32.0 %
Disodium alpha-glycerophosphate max. 4.0 %
Phosphatase inhibition test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR