3,6-Di-tert-butylcarbazole ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3,6-Di-tert-butyl-9H-carbazole
D3952-1G
D3952-5G
TCIAD3952-1GEA
31
EUR
TCIAD3952-1G
TCIAD3952-5G
3,6-Di-tert-butylcarbazole ≥98.0% (by GC)
3,6-Di-tert-butylcarbazole
Formula:
C₂₀H₂₅N Peso molecolare: 279,43 g/mol |
MDL Number:
MFCD03425849 CAS Number: 37500-95-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 230.0 to 234.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...