3,5-Dimethyl-1H-pyrrole-2-carbaldehyde ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
D3950-1G
D3950-5G
TCIAD3950-1GEA
85
EUR
TCIAD3950-1G
TCIAD3950-5G
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde ≥98.0% (by GC)
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde
Formula:
C7H9NO |
MDL Number:
MFCD00111522 CAS Number: 2199-58-8 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 89.0 to 93.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...