Stampa…

3,3'-Dibromo-5,5'-bis(trimethylsilyl)-2,2'-bithiophene ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,3-Dibromo-5,5-bis(trimethylsilyl)-2,2-bithiophene

D3941-1G D3941-5G
TCIAD3941-1GEA 104 EUR
TCIAD3941-1G TCIAD3941-5G
3,3'-Dibromo-5,5'-bis(trimethylsilyl)-2,2'-bithiophene ≥98.0% (by GC)
3,3'-Dibromo-5,5'-bis(trimethylsilyl)-2,2'-bithiophene
Formula: C₁₄H₂₀Br₂S₂Si₂
Peso molecolare: 468,41 g/mol
MDL Number: MFCD16621088
CAS Number: 207742-50-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 91.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR