Stampa…

3,4-Dibromo-1H-pyrrole-2,5-dione ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 3,4-Dibromomaleimide

D3910-1G D3910-5G
TCIAD3910-1GEA 58 EUR
TCIAD3910-1G TCIAD3910-5G
3,4-Dibromo-1H-pyrrole-2,5-dione ≥98.0% (by HPLC, titration analysis)
3,4-Dibromo-1H-pyrrole-2,5-dione
Formula: C4HBr2NO2
Peso molecolare: 254,8600 g/mol
MDL Number: MFCD00185696
CAS Number: 1122-10-7
UN: 1759
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR