Stampa…

trans-2,5-Dichlorocinnamic acid ≥96.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: trans-3-(2,5-Dichlorophenyl)-2-propenoic Acid

D3794-25G D3794-5G
TCIAD3794-25GEA 149 EUR
TCIAD3794-25G TCIAD3794-5G
trans-2,5-Dichlorocinnamic acid ≥96.0% (by GC, titration analysis)
trans-2,5-Dichlorocinnamic acid
Formula: C₉H₆Cl₂O₂
Peso molecolare: 217,05 g/mol
MDL Number: MFCD03002786
CAS Number: 20595-47-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 190.0 to 194.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR