Desloratadine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
Synonyms:
8-Chloro-11-(piperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
Formula:
C₁₉H₁₉ClN₂ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00871949 CAS Number: 100643-71-8 Merck Index: 14,02922 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 155.0 to 159.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...