2,6-Dichlorodiphenylamine ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
D3778-25G
D3778-250G
TCIAD3778-25GEA
26
EUR
TCIAD3778-25G
TCIAD3778-250G
2,6-Dichlorodiphenylamine ≥98.0% (by GC)
2,6-Dichlorodiphenylamine
Formula:
C₁₂H₉Cl₂N |
MDL Number:
MFCD00269648 CAS Number: 15307-93-4 EINECS: 239-349-0 |
Risultati del test di specificazione
Appearance | White to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 49.0 to 53.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...