Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
|
Attenzione
|
D3775-1G
D3775-25G
D3775-5G
TCIAD3775-1GEA
16
EUR
TCIAD3775-1G
TCIAD3775-25G
TCIAD3775-5G
Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate ≥98.0% (by HPLC, total nitrogen)
Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate
Formula:
C₁₃H₁₉NO₄ |
MDL Number:
MFCD00005951 CAS Number: 1149-23-1 EINECS: 214-561-6 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...